Order Entry
Export
ContactUsLinkComponent
Diethyl pyrazole-3,5-dicarboxylate 99%
Irritant
Diethyl pyrazole-3,5-dicarboxylate 99%
Catalog # ACRO296520050
CAS Number:  
undefined
Irritant
Diethyl pyrazole-3,5-dicarboxylate 99%
Catalog # ACRO296520050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    51 to 58 °C
  • GC:
    ≥98.5 %

Specification Test Results

About this item

 :
Melting Pt:51…55 °C
 :212.21 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:37687-24-4
Formula:C₉H₁₂N₂O₄
MDL Number:MFCD00152167