Order Entry
Export
ContactUsLinkComponent
2,5-Dibromo-3,4-dinitrothiophene
Irritant
2,5-Dibromo-3,4-dinitrothiophene
Catalog # ACRO278220050
CAS Number:  
undefined
Irritant
2,5-Dibromo-3,4-dinitrothiophene
Catalog # ACRO278220050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    135 to 140 °C
  • GC:
    ≥94 %

Specification Test Results

About this item

 :
 :331.94 g/mol
Melting Pt:132…134 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:52431-30-8
Formula:C₄Br₂N₂O₄S
MDL Number:MFCD00015537