
2,5-Dihydrofuran 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1708-29-8
:
Formula:C₄H₆O
MDL Number:MFCD00003219
:70.09 g/mol
Boiling Pt:66…67 °C (760mbar)
Density:0.92 g/cm³
Flash Pt:–016 °C
Storage Temperature:Ambient
UN:3271
ADR:3,II
:GHS02
Specification Test Results
- Appearance:Colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4310 to 1.4330 (20°C, 589 nm)
- GC:≥97.5 %