Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II) carbonate ACS

Lead(II) carbonate ACS

Health
Irritant
Environment
Lead(II) carbonate ACS
Certificates
Lead(II) carbonate ACS

2 Options Available Below

Product Details & Documents
Synonyms:  Lead carbonat
  • CAS Number:598-63-0
  • Formula:PbCO₃
  • MDL Number:MFCD00064631
  • Molecular Weight:267.21 g/mol
  • Melting Pt:400 °C
  • Density:6,43 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • UN:3077
  • ADR:9,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Powder
  • Chloride (Cl):
    ≤0.002 %
  • Nitrate and Nitrite (as NO3):
    (limit about 0.005%) Passes test
  • Cadmium (Cd):
    ≤0.002 %
  • Calcium (Ca):
    ≤0.01 %
  • Iron (Fe):
    ≤0.005 %
  • Potassium (K):
    ≤0.02 %
  • Sodium (Na):
    ≤0.05 %
  • Zinc (Zn):
    ≤0.003 %
  • Insoluble matter:
    ≤0.02 % (in dilute acetic acid)
Product Family Options

Product Information