Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bis-trispropane 99%

Bis-trispropane 99%
Certificates
Bis-trispropane 99%

4 Options Available Below

Product Details & Documents
Synonyms:  1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
  • CAS Number:64431-96-5
  • Formula:C₁₁H₂₆N₂O₆
  • MDL Number:MFCD00004689
  • Molecular Weight:282.34 g/mol
  • Melting Pt:164…165 °C
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    161 to 168 °C
  • Loss on drying:
    ≤1 %
  • Solubility:
    (0.1 M in water) Clear colorless
  • UV:
    (0.1 M in water)
  • Titration with HClO4:
    98.5 to 101.5 %
Product Family Options

Product Information