Order Entry
Export
ContactUsLinkComponent
 
2,2-Diphenylethanol 97%
 
undefined
2,2-Diphenylethanol 97%

 

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    57 to 61 °C
  • GC:
    ≥96.0 %