

α,α'-Dibromo-m-xylene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:626-15-3
:
Formula:C₈H₈Br₂
MDL Number:MFCD00000178
:263.96 g/mol
Boiling Pt:135…140 °C (20 mmHg)
Melting Pt:75…77 °C
Density:1,959 g/cm³ (0 °C)
Storage Temperature:Ambient
UN:2928
ADR:6.1,II
:GHS05
:GHS07
Specification Test Results
- Appearance:White to brown Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:74 to 77 °C
- HPLC:≥96.0 %
- NMR (1H-NMR):Consistent with structure