Order Entry
Export
ContactUsLinkComponent
4,5-Diaminopyrimidine 97%
Irritant
4,5-Diaminopyrimidine 97%
Catalog # ACRO112400010
CAS Number:  
undefined
Irritant
4,5-Diaminopyrimidine 97%
Catalog # ACRO112400010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Brown Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %
  • GC:
    ≥97.5 %
  • Specific optical rotation:
    +53° to +56° (22°C, 589 nm) (c=1, 1,2-dichlorethane)

Specification Test Results

About this item

 :
 :110.1200 g/mol
Boiling Pt:229 °C (32 mmHg)
Melting Pt:200 °C
ADR:6.1,III
 :GHS07
Storage Temperature:Ambient
CAS Number:13754-19-3
Formula:C₄H₆N₄
MDL Number:MFCD00006113