Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4′-Hydroxy-2-phenylacetophenone
4′-Hydroxy-2-phenylacetophenone 97%
4′-Hydroxy-2-phenylacetophenone 97%
:
Thermo Fisher Scientific
4′-Hydroxy-2-phenylacetophenone 97%
:
Thermo Fisher Scientific
:
2491-32-9
:
:
C₁₄H₁₂O₂
:
MFCD00002360
:
212.24 g/mol
:
148…151 °C
:
GHS07
:
Ambient
Appearance:
Cream Crystalline powder
Infrared spectrum:
Conforms
Melting point:
147 to 151 °C
HPLC:
≥96.0 %
ACRO10598