Orders LinkContactUsLinkComponent
Accessories for microcarriers
   Sustainable Options Available
Supplier:   Corning
Accessories for microcarriers
Supplier:  Corning
   Sustainable Options Available
Certificates
Accessories for microcarriers
4 Options Available Below
Product Details & Documents
These polypropylene aseptic transfer screw caps prevents adventitious contamination in for microcarriers bottles during bioreactor loading.
  • Chemically resistant and heat sealable flexible tubing
  • Tubing length with connector 30" (76,2 cm)
  • Disposable and sterile (10⁻⁶ SAL)
 : These caps are individually double bagged.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-2919
  • 734-2920
  • 734-2921
  • 734-2922
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 734-2919
    Aseptic transfer cap, PP, orange, disposable ,45 mm, male luer lock, for 100 g microcarrier bottle
    Aseptic transfer cap, PP, orange, disposable ,45 mm, male luer lock, for 100 g microcarrier bottle
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 226,00 €
    226,00 €
    1 * 1 items
       226,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2919
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
      Low Manufacturing Impact
  • 734-2920
    Aseptic transfer cap, PP, orange, disposable ,45 mm, MPC, for 100 g microcarrier bottle
    Aseptic transfer cap, PP, orange, disposable ,45 mm, MPC, for 100 g microcarrier bottle
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 242,00 €
    242,00 €
    1 * 1 items
       242,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2920
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
      Low Manufacturing Impact
  • 734-2921
    Aseptic transfer cap, PP, orange, disposable ,48 mm, male luer lock, for 500 g microcarrier bottle
    Aseptic transfer cap, PP, orange, disposable ,48 mm, male luer lock, for 500 g microcarrier bottle
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 213,00 €
    213,00 €
    1 * 1 items
       213,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2921
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
      Low Manufacturing Impact
  • 734-2922
    Aseptic transfer cap, PP, orange, disposable ,48 mm, MPC, for 500 g microcarrier bottle
    Aseptic transfer cap, PP, orange, disposable ,48 mm, MPC, for 500 g microcarrier bottle
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 228,00 €
    228,00 €
    1 * 1 items
       228,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2922
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
      Low Manufacturing Impact