Dimethyl sulphoxide, anhydrous ≥99.9%
Catalog # 90082.
Supplier: Biotium
CAS Number:
Specifications
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- Color & Form:Colorless liquid
- Purity:≥ 99.9%
- Water:Max 0.005%
- Residual on evaporation:Max 0.0005%
Specification Test Results
About this item
:
:78.14 g/mol
Storage Temperature:Ambient
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089