Storage vials, depyrogenated glass
Catalog # EPSC23-CTP4/PF
Supplier: EP SCIENTIFIC
Specifications
- Cap included:Yes
- Cap material:Urea
- Cap Size:13-425
- Ø×H:15×45 mm
- Capacity:4,00 ml
- Colour:Clear
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:NAR Certificate of Retirement
- Pk:72
Specifications
About this item
Clear glass with urea closure.
- Packaging and storing articles that will be terminally sterilized
- Storage of laboratory reagents and medias
- Sample storage and water sampling
- Meet endotoxin levels of less than 0,06 EU/ml
Delivery: It is supplied with certificate of analysis.