Order Entry
Export
ContactUsLinkComponent
3,5-Difluorophenylmagnesium bromide 0.5 M in THF, AcroSeal®
 
 
 
 
3,5-Difluorophenylmagnesium bromide 0.5 M in THF, AcroSeal®
  427441000.
 :  
undefined
 
 
 
 
3,5-Difluorophenylmagnesium bromide 0.5 M in THF, AcroSeal®
  427441000.
 :  

 

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Appearance:
    Yellow to green or brown Solution
  • Titration Grignard:
    0.40 to 0.75 M
  • Refractive index:
    1.5610 to 1.5630 (20°C, 589 nm)
  • GC:
    ≥97.5 %

 

 

 :
 :217.296 g/mol
 :65 °C (760 mmHg)
 :0.971 g/cm³
 :–17 °C
 :Ambient
 :2924
 :3,II
 :GHS07
 :GHS05
 :GHS02
 :GHS08
 :62351-47-7
 :C₆H₃BrF₂Mg
 :MFCD00672005