Order Entry
Export
ContactUsLinkComponent
 
Chloro(diisopropyl)phosphine 96%, AcroSeal®
  427270500.
 :  
undefined
Chloro(diisopropyl)phosphine 96%, AcroSeal®
  427270500.
 :  

 

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    50 mL
  • Seq:
    2370

 

 

  • Appearance:
    Clear light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4740 to 1.4760 (20°C, 589 nm)
  • GC:
    ≥95 %
  • Residue after evaporation:
    ≤0.002 %
  • Acidity/alkalinity:
    ≤0.0001 meq/g
  • Calcium (Ca):
    ≤0.5 ppm
  • Copper (Cu):
    ≤0.05 ppm
  • Iron (Fe):
    ≤0.2 ppm
  • Lead (Pb):
    ≤0.05 ppm
  • Magnesium (Mg):
    ≤0.1 ppm
  • Potassium (K):
    ≤0.2 ppm
  • Sodium (Na):
    ≤1 ppm
  • Zinc (Zn):
    ≤0.1 ppm
  • Total phosphorus (P):
    ≤0.5 ppm
  • Total silicon (Si):
    ≤0.02 ppm
  • Total sulfur (S):
    ≤5 ppm

 

 

 :
 :152.6 g/mol
 :69 °C (33 mmHg)
 :3 °C
 :Ambient
 :2924
 :3,II
 :GHS05
 :GHS02
 :40244-90-4
 :C₆H₁₄ClP
 :MFCD00015027