
N-Carbobenzoxy-1,3-diaminopropane hydrochloride 98%
:17400-34-9
:
:C₁₁H₁₆N₂O₂
:MFCD00270153
:244.72 g/mol
:185…189 °C
:GHS07
:Ambient
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:185 to 189 °C
- NMR:consistent with structure
- Titration with HClO4:≥97.5 %