
1,4-Diiodoperfluorobutane 98%
ACRO426320050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Clear colorless to light yellow or pink Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4280 to 1.4320 (20°C, 589 nm)
- GC:≥97.5 %
:
:453.84 g/mol
:150 °C (1013 hPa)
:–9 °C
:2.474 g/cm³
:GHS07
:Refrigerator
:375-50-8
:C₄F₈I₂
:MFCD00042263