
1,7-Phenanthroline 99%
Catalog # ACRO426110010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:70 to 79 °C
- GC:≥98.5 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:180.21 g/mol
Boiling Pt:360 °C (1013 hPa)
Melting Pt:77…78 °C
Density:1,25 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:230-46-6
Formula:C₁₂H₈N₂
MDL Number:MFCD00004971