
4,7-Phenanthroline 98%
Catalog # ACRO426100010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:170 to 174 °C
- HPLC:≥97.5 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:180.21 g/mol
Melting Pt:172…174 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:230-07-9
Formula:C₁₂H₈N₂
MDL Number:MFCD00004987