
2,3-Diaminophenol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:59649-56-8
:
Formula:C₆H₈N₂O
MDL Number:MFCD00075199
Melting Pt:161…165 °C
:124.14 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light brown to dark brown Powder or crystalline powder
- Infrared spectrum:Conforms
- Melting point:161 to 165 °C
- HPLC:≥96.0 %