
2,2-Dimethyl-1,3-dioxolane-4-methanamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22195-47-7
:
Formula:C₆H₁₃NO₂
MDL Number:MFCD01321384
:131.175 g/mol
Boiling Pt:164…165 °C (760 mmHg)
Density:1.012 g/cm³
Flash Pt:68 °C
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4370 to 1.4390 (20°C, 589 nm)
- GC:≥96.0 %