2,2'-Thenil 98+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 1,2-Di(thiophen-2-yl)ethane-1,2-dione
- CAS Number:7333-07-5
- Formula:C₁₀H₆O₂S₂
- MDL Number:MFCD00173678
- Molecular Weight:222.29 g/mol
- Melting Pt:81…84 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Synonyms: 1,2-Di(thiophen-2-yl)ethane-1,2-dione
- CAS Number:7333-07-5
- Formula:C₁₀H₆O₂S₂
- MDL Number:MFCD00173678
- Molecular Weight:222.29 g/mol
- Melting Pt:81…84 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III
Specifications for Product Family
Specification Test Results
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Melting point:81 to 84 °C
- GC:≥98.0 %



