Order Entry
Export
ContactUsLinkComponent
(R)-(+)-1,2-Epoxypropane 98+%
(R)-(+)-1,2-Epoxypropane 98+%
Certificates
undefined
(R)-(+)-1,2-Epoxypropane 98+%

Specification Test Results

  • Appearance:
    Colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3650 to 1.3680 (20°C, 589 nm)
  • GC:
    ≥98.0 %
  • Specific optical rotation:
    +12° to +15° (20°C, 589 nm) (Neat)

Specification Test Results

Related Information