Order Entry
Export
ContactUsLinkComponent
tert-Butyl-1,4-diazepane-1-carboxylate 98%
Corrosive
tert-Butyl-1,4-diazepane-1-carboxylate 98%
Catalog # ACRO399530050
CAS Number:  
undefined
Corrosive
tert-Butyl-1,4-diazepane-1-carboxylate 98%
Catalog # ACRO399530050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥97.5 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

About this item

 :
 :200.28 g/mol
Boiling Pt:92…96 °C (.5 mmHg)
Density:1.029 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:2735
ADR:8,III
 :GHS05
CAS Number:112275-50-0
Formula:C₁₀H₂₀N₂O₂
MDL Number:MFCD00276987