



3,4-Difluorophenylmagnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 399511000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Yellow to dark brown Solution
- Titration Grignard:0.40 to 0.60 M
Specification Test Results
About this item
:
Boiling Pt:65 °C (760 mmHg)
Density:0.965 g/cm³
Flash Pt:–17 °C
ADR:3,II
:217.296 g/mol
UN:2924
:GHS07
:GHS05
:GHS02
:GHS08
Storage Temperature:Ambient
CAS Number:90897-92-0
Formula:C₆H₃BrF₂Mg
MDL Number:MFCD00672004