
2-Bromopyrimidine 97+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4595-60-2
:
Formula:C₄H₃BrN₂
MDL Number:MFCD00014601
Boiling Pt:62…64 °C (1.5 mmHg)
Melting Pt:55…56 °C
Density:Min, 1 g/cm³ (20 °C)
:158.99 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:55 to 59 °C
- HPLC:≥97.0 %
- Specific optical rotation:-11° to -16° (20 °C, 589 nm) (c=4, MeOH)