Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Iodopyridine 96%

Irritant
4-Iodopyridine 96%
Certificates
4-Iodopyridine 96%

2 Options Available Below

Product Details & Documents
  • CAS Number:15854-87-2
  • Formula:C₅H₄IN
  • MDL Number:MFCD02093937
  • Molecular Weight:205 g/mol
  • Melting Pt:92…96 °C
  • Storage Temperature:Freezer

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Light brown to dark green-brown Crystals or powder (May darken on storage)
  • Infrared spectrum:
    Conforms
  • GC:
    ≥95 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 43,80 €
    43,80 €
    1 * 1 g
       43,80 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 181,00 €
    181,00 €
    1 * 5 g
       181,00 €