
2-Fluoro-4-(trifluoromethyl)benzoyl chloride 97%
Catalog # ACRO397820050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4650 to 1.4670 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
About this item
:
:226.56 g/mol
Boiling Pt:188…189 °C (1013 hPa)
Density:1.5 g/cm³
Flash Pt:81 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
CAS Number:126917-10-0
Formula:C₈H₃ClF₄O
MDL Number:MFCD00061155