
2,5-Dibromothiazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4175-78-4
:
Formula:C₃HBr₂NS
MDL Number:MFCD00016891
:242.92 g/mol
Melting Pt:45…49 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:45 to 50 °C
- GC:≥96.0 %