Lead(II) chloride 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Lead dichloride
- CAS Number:7758-95-4
- Formula:Cl₂Pb
- MDL Number:MFCD00011157
- Molecular Weight:278.11 g/mol
- Boiling Pt:950 °C (1013 hPa)
- Melting Pt:501 °C
- Density:5,85 g/cm³ (20 °C)
- Storage Temperature:Ambient
- UN:2291
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Synonyms: Lead dichloride
- CAS Number:7758-95-4
- Formula:Cl₂Pb
- MDL Number:MFCD00011157
- Molecular Weight:278.11 g/mol
- Boiling Pt:950 °C (1013 hPa)
- Melting Pt:501 °C
- Density:5,85 g/cm³ (20 °C)
- Storage Temperature:Ambient
- UN:2291
- ADR:6.1,III
Documents
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Titration Complexometric:≥98.5 %



