1,1-Diethoxybutane 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3658-95-5
:
Formula:C₈H₁₈O₂
MDL Number:MFCD00038316
:146.23 g/mol
Boiling Pt:143 °C (760 mmHg)
Density:0.83 g/cm³
Flash Pt:30 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3950 to 1.3980 (20°C, 589 nm)
- GC:≥96.0 %