5,5'-Dimethyl-2,2'-bithiophene 99%
ACRO357950010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:Yellow to light brown Crystals
- Infrared spectrum:Conforms
- Melting point:65 to 68 °C
- GC:≥99.9 %
:
:194.31 g/mol
:65…68 °C
:Refrigerator
:16303-58-5
:C₁₀H₁₀S₂
:MFCD01927313