Order Entry
Export
Orders LinkContactUsLinkComponent
Dimethyl-L-aspartate hydrochloride 98%

Dimethyl-L-aspartate hydrochloride 98%

Dimethyl-L-aspartate hydrochloride 98%
Certificates
Dimethyl-L-aspartate hydrochloride 98%

2 Options Available Below

Product Details & Documents

Appearance: White to off-white Crystalline powder

Synonyms:  (S)-Aminosuccinic Acid Dimethyl Ester Hydrochloride, L-Asparaginic Acid Dimethyl Ester Hydrochloride, Dimethyl (S)-Aminosuccinate Hydrochloride, H-Asp(ome)-OMe HCl, Dimethyl L-Asparaginate Hydrochloride, H-Asp(OMe)-OMe.HCl, L-Aspartic Acid Dimethyl Ester Hydrochloride
  • CAS Number:32213-95-9
  • Formula:C₆H₁₁NO₄
  • MDL Number:MFCD00038878
  • Molecular Weight:197.62 g/mol
  • Melting Pt:115…117 °C
  • Storage Temperature:Ambient

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥97.5 %
  • Specific optical rotation:
    +13° to +17° (20°C, 589 nm) (c=1, CH3OH)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 68,20 €
    68,20 €
    1 * 5 g
       68,20 €

  • Plastic bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 250,00 €
    250,00 €
    1 * 25 g
       250,00 €