
1,1'-Sulphonyldiimidazole 98+%
Catalog # ACRO374780050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Cream Crystalline powder
- Infrared spectrum:Conforms
- Melting point:136 to 139 °C
- HPLC:≥98.0 %
Specification Test Results
About this item
:
:198.20 g/mol
Melting Pt:135…137 °C
:GHS07
Storage Temperature:Ambient
CAS Number:7189-69-7
Formula:C₆H₆N₄O₂S
MDL Number:MFCD00015893