Order Entry
Export
ContactUsLinkComponent
tert-Butyl peroxyacetate 50% in aromatic free mineral spirit
Flammable
Toxic
Health
tert-Butyl peroxyacetate 50% in aromatic free mineral spirit
Catalog # ACRO349862500
CAS Number:  
undefined
Flammable
Toxic
Health
tert-Butyl peroxyacetate 50% in aromatic free mineral spirit
Catalog # ACRO349862500
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    Clear solution
  • Titration Iodometric:
    49 to 51 %
  • Color scale:
    ≤20 APHA
  • Hydrolyzable chloride:
    ≤50 ppm Inorganic + organic
  • Impurity:
    ≤1000 ppm Hydroperoxides (as TBHP)

Specification Test Results

About this item

 :
 :132.159 g/mol
Melting Pt:–20 °C
Density:0.82 g/cm³
Flash Pt:43 °C
Storage Temperature:Refrigerator
UN:3103
ADR:5.2,
 :GHS02
 :GHS06
 :GHS08
CAS Number:107-71-1
Formula:C₆H₁₂O₃
MDL Number:MFCD00048240