
2-(tert-Butoxycarbonylamino)-1-ethanol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:26690-80-2
:
Formula:C₇H₁₅NO₃
MDL Number:MFCD00056657
:161.20 g/mol
Density:1.042 g/cm³
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4485 to 1.4505 (20°C, 589 nm)
- GC:≥97.5 %