
2,4-Dihydroxy-6-methylpyridine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3749-51-7
:
Formula:C₆H₇NO₂
MDL Number:MFCD01860849
:125.13 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Beige Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %