
7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline 98%
Catalog # ACRO277822500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:Yellow Powder and/or flakes
- Infrared spectrum:Conforms
- Melting point:187 to 191 °C
- GC:≥97.5 %
Specification Test Results
About this item
:
Melting Pt:187…191 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:177.203 g/mol
:GHS07
CAS Number:4602-73-7
Formula:C₁₀H₁₁NO₂
MDL Number:MFCD00143517