


Sodium percarbonate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15630-89-4
:
Formula:2Na₂CO₃·3H₂O₂
MDL Number:MFCD00043204
:314.017 g/mol
Storage Temperature:Ambient
UN:3378
ADR:5.1,III
:GHS07
:GHS05
:GHS03
Specification Test Results
- Appearance:White Granular powder or granules
- Titration with KMnO4:12.5 to 14.5 % (Available Oxygen)