
3-Oxabicyclo[3.1.0]hexane-2,4-dione 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5617-74-3
:
Formula:C5H4O3
MDL Number:MFCD00126929
:112.08 g/mol
Boiling Pt:100…102 °C
Melting Pt:54…60 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:54 to 60 °C
- GC:≥97.5 %