
N-Boc-L-prolinal 97%
:69610-41-9
:
:C₁₀H₁₇NO₃
:MFCD00274186
:199.25 g/mol
:211 °C (760 mmHg)
:1.063 g/cm³
:GHS07
:Freezer
- Appearance:Clear colorless to light yellow to orange Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4590 to 1.4630 (20°C, 589 nm)
- GC:≥96.0 %
- Specific optical rotation:-90° to -105° (20°C, 589 nm) (c=1, CHCl3)