Order Entry
Export
ContactUsLinkComponent
 
Aluminium oxide neutral Brockmann Grade I 40-300 µm, 60A for chromatography
 
undefined
Aluminium oxide neutral Brockmann Grade I 40-300 µm, 60A for chromatography
 

 

  • Appearance:
    White to off-white Powder
  • Particle size:
    ≤5 % (<40 µm)
  • Specific surface area:
    185 to 225 m²/g
  • pH:
    6.5 to 7.5 (10% aqueous suspension)
  • Sodium oxide (Na2O):
    ≤0.4 %
  • Silicon dioxide (SiO2):
    ≤0.03 %
  • Iron(III)oxide (Fe2O3):
    ≤0.03 %