Order Entry
Export
ContactUsLinkComponent
Diglycidyl ether 95%
Toxic
Corrosive
Diglycidyl ether 95%
Certificates
undefined
Toxic
Corrosive
Diglycidyl ether 95%

Specification Test Results

  • Appearance:
    Clear colorless to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4460 to 1.4500 (20°C, 589 nm)
  • GC:
    ≥94 %

Specification Test Results

Related Information