0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Procainamide hydrochloride
Procainamide hydrochloride 99%
Procainamide hydrochloride 99%
:
Thermo Fisher Scientific
Procainamide hydrochloride 99%
:
Thermo Fisher Scientific
:
614-39-1
:
:
C₁₃H₂₂ClN₃O
:
MFCD00012998
:
271.789 g/mol
:
166…170 °C
:
GHS07
:
Ambient
Appearance:
White to light yellow Powder
Infrared spectrum:
Conforms
Melting point:
166 to 170 °C
Titration with HClO4:
≥98.5 %
ACRO20730