Order Entry
Export
ContactUsLinkComponent
 
 
(-)-Perillaldehyde 90%
 
undefined
 
(-)-Perillaldehyde 90%

 

  • Appearance:
    Clear light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5040 to 1.5100 (20°C, 589 nm)
  • Assay:
    ≥88 %
  • Specific gravity:
    (20/20°C) 0.965 to 0.975
  • Specific optical rotation:
    -105° to -125° (20°C, 589 nm) (c=10, C2H5OH)