
Disperse orange 3 90%, pure
Catalog # ACRO201620500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Appearance:Red to dark brown Powder
- Infrared spectrum:Conforms
- TLC:≥88 %
Specification Test Results
About this item
CAS Number:730-40-5
:
Formula:C₁₂H₁₀N₄O₂
MDL Number:MFCD00007856
:242.238 g/mol
Melting Pt:200 °C
:GHS07
Storage Temperature:Ambient