Order Entry
Export
Orders LinkContactUsLinkComponent
Fluorescamine, pure

Fluorescamine, pure

Fluorescamine, pure
Certificates
Fluorescamine, pure

4 Options Available Below

Product Details & Documents

CAS No.: 38183-12-9

Synonyms:  Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione
  • CAS Number:38183-12-9
  • Formula:C₁₇H₁₀O₄
  • MDL Number:MFCD00005928
  • Molecular Weight:278.26 g/mol
  • Melting Pt:155…157 °C
  • Storage Temperature:Ambient

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Light yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    155 to 157 °C
  • Specific extinction E (1%/1cm):
    ≥900 (235 nm, ether)
  • Suitability:
    fluorescence ind. for prim. amines and amino acids
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 56,10 €
    56,10 €
    1 * 25 mg
       56,10 €

  • Glass bottle
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 138,00 €
    138,00 €
    1 * 100 mg
       138,00 €

  • Glass bottle
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 467,00 €
    467,00 €
    1 * 500 mg
       467,00 €

  • Glass bottle
    2,5 g
    Log in for availability
    Log in for availability
    1 * 2,5 g
    1 * 2,5 g - 1 770,00 €
    1 770,00 €
    1 * 2,5 g
       1 770,00 €