Aluminium oxide basic Brockmann Grade I 40-300 µm, 60A for chromatography
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1344-28-1
:
Formula:Al₂O₃
MDL Number:MFCD00003424
:101.96 g/mol
Boiling Pt:2977 °C (760 mmHg)
Melting Pt:2030 °C
Density:3.97 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder
- Particle size:≤5 % (<40 µm)
- Specific surface area:185 to 225 m²/g
- pH:8.5 to 10.5 (10% aqueous suspension)
- Sodium oxide (Na2O):≤0.4 %
- Silicon dioxide (SiO2):≤0.03 %
- Iron(III)oxide (Fe2O3):≤0.03 %