2-(3,4-Dimethoxyphenyl)ethanol 98%
Catalog # ACRO189000100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to light yellow Powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:44 to 48 °C
- GC:≥97.5 %
- UV:at 270 nm A: ≤0.1
- Titration with NaOH:≤99.75 %
Specification Test Results
About this item
:
:182.22 g/mol
Boiling Pt:172…174 °C (17 mmHg)
Melting Pt:44…48 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
CAS Number:7417-21-2
Formula:C₁₀H₁₄O₃
MDL Number:MFCD00002894