
1,2-Dipiperidinoethane 98%
:1932-04-3
:
:C₁₂H₂₄N₂
:MFCD00014644
:196.338 g/mol
:265 °C (760 mmHg)
:1….5 °C
:0.91 g/cm³
:110 °C
:GHS07
:Ambient
- Appearance:Clear yellow to amber to orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4865 to 1.4885 (20°C, 589 nm)
- GC:≥97.5 %
- Specific optical rotation:-36.5° to -40° (20°C, 589 nm) (1 % in 0.1M NaOH)
- Sulfate (SO4):≤0.1 %