Order Entry
Export
ContactUsLinkComponent
 
Polyethylene glycol, M.W. 600,000 - 1,000,000
 
undefined
Polyethylene glycol, M.W. 600,000 - 1,000,000

 

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤2 % (10 g, 5 h, 10 mmHg)
  • Viscosity:
    300 to 600 mPa.s (2 % w/w in water at 25°C)